Merck
  • UV-Vis, fluorescence and NMR spectroscopic investigations on inclusion properties of a designed tetrahomocalix[8]arene with fullerenes C60 and C70 in solution.
UV-Vis, fluorescence and NMR spectroscopic investigations on inclusion properties of a designed tetrahomocalix[8]arene with fullerenes C60 and C70 in solution.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-10-04)
Amal Halder, Suchitra Bhatt, Sandip K Nayak, Subrata Chattopadhyay, Sumanta Bhattacharya
ABSTRAKT
The present article reports the spectroscopic investigations on non-covalent interaction of fullerenes C(60) and C(70) with a macrocyclic receptor molecule, namely, 1,3,5,7-tetrahomo-p-tert-butylcalix[8]arene (1) in toluene. Jobs method of continuous variation reveals 1:1 stoichiometry for the fullerene complexes of 1. The most fascinating feature of the present study is that 1 binds selectively C(60) compared to C(70) as obtained from binding constant (K) data of C(60)-1 (K(C60-1)) and C(70)-1 (K(C70-1)) complexes which are enumerated to be 265,000 dm(3) mol(-1) and 63,43 dm(3) mol(-1), respectively, and selectivity in binding (K(C60-1)/K(C70-1)) is estimated to be 4.18 as obtained from UV-Vis study. Steady state fluorescence studies reveal quenching of fluorescence of 1 in presence of fullerenes and the K value of the C(60)-1 and C(70)-1 complexes are estimated to be 80,760 and 68,780 dm(3) mol(-1), respectively, with selectivity in binding (K(C60-1)/K(C70-1)) ~1.18. (1)H NMR analysis provides very good support in favor of strong binding between C(60) and 1. The high value of K value for C(60)-1 complex indicates that 1 forms an inclusion complex with C(60).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
[5,6]-Fullerene-C70, 98%
Sigma-Aldrich
[5,6]-Fullerene-C70, sublimed, ≥99% (HPLC)