Merck
DFT studies of COOH tip-functionalized zigzag and armchair single wall carbon nanotubes.
Journal of molecular modeling (2011-10-04)
Elżbieta Chełmecka, Karol Pasterny, Teobald Kupka, Leszek Stobiński
ABSTRAKT
Structure and energy calculations of pristine and COOH-modified model single wall carbon nanotubes (SWCNTs) of different length were performed at B3LYP/6-31G* level of theory. From 1 to 9 COOH groups were added at the end of the nanotube. The differences in structure and energetics of partially and fully functionalized SWCNTs at one end of the nanotube are observed. Up to nine COOH groups could be added at one end of (9,0) zigzag SWCNT in case of full functionalization. However, for (5,5) armchair SWCNT, the full functionalization was impossible due to steric crowding and rim deformation. The dependence of substituent attachment energy on the number of substituents at the carbon nanotube rim was observed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
9-Anthracenecarboxylic acid, 99%