Merck
Meet me on the other side: trans-bilayer modulation of a model voltage-gated ion channel activity by membrane electrostatics asymmetry.
PloS one (2011-10-08)
Loredana Mereuta, Alina Asandei, Tudor Luchian
ABSTRAKT
While it is accepted that biomembrane asymmetry is generated by proteins and phospholipids distribution, little is known about how electric changes manifested in a monolayer influence functional properties of proteins localized on the opposite leaflet. Herein we used single-molecule electrophysiology and investigated how asymmetric changes in the electrostatics of an artificial lipid membrane monolayer, generated oppositely from where alamethicin--a model voltage-gated ion channel--was added, altered peptide activity. We found that phlorizin, a membrane dipole potential lowering amphiphile, augmented alamethicin activity and transport features, whereas the opposite occurred with RH-421, which enhances the monolayer dipole potential. Further, the monolayer surface potential was decreased via adsorption of sodium dodecyl sulfate, and demonstrated that vectorial modification of it also affected the alamethicin activity in a predictive manner. A new paradigm is suggested according to which asymmetric changes in the monolayer dipole and surface potential extend their effects spatially by altering the intramembrane potential, whose gradient is sensed by distantly located peptides.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Alamethicin, Ready Made Solution from Trichoderma viride, 5 mg/mL in DMSO
Sigma-Aldrich
Alamethicin from Trichoderma viride, ≥98% (HPLC)