Merck
Novel screen-printed electrode for the determination of dodecyltrimethylammonium bromide in water samples.
Drug testing and analysis (2011-10-15)
Gehad G Mohamed, Tamer Awad Ali, M F El-Shahat, M A Migahed, A M Al-Sabagh
ABSTRAKT
The construction and electrochemical response characteristics of a screen-printed electrode (SPE) for the determination of dodecyltrimethylammonium bromide (DTAB) are described. The sensor was based on the use of DTA-tetraphenylborate ion association complex as an electroactive material in screen-printed electrode with dioctylsebacate (DOS) as a solvent mediator. In aqueous solution of pH 3, the sensor displayed a stable response for six months with reproducible potential and linear response for surfactant over the concentration range 1.20 × 10(-2) -5.6 × 10(-7) mol L(-1) at 25 °C with Nernstian slope of 55.95 ± 0.58 mV decade(-1) for detection limit of 6.8 x 10(-6) mol L(-1) . The response time was 6-10 s. The selectivity coefficients indicate excellent selectivity for DTAB over many common cations (e.g. Mg(2+), Na(+), K+, Co(2+), Ni(2+), Ca(2+), Cl(-), I(-), SO(4)(-2) and cetylpyridinium chloride (CPC). The sensor was used successfully for the determining of DTAB in pure form and water samples with average recoveries of 99.98, 98.78, and 99.99%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium tetraphenylborate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraphenylborate, puriss. p.a., ACS reagent, ≥99.5% (NT)
Sigma-Aldrich
Ammonium tetraphenylborate, 99%
Supelco
Dodecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥98.5% (AT)
Sigma-Aldrich
Dodecyltrimethylammonium bromide, BioXtra, ~99%
Sigma-Aldrich
Dodecyltrimethylammonium chloride, ≥99.0% (AT)
Sigma-Aldrich
Dodecyltrimethylammonium chloride, purum, ≥98.0% anhydrous basis (AT)
Sigma-Aldrich
Dodecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Potassium tetraphenylborate, 97%