Merck
Chemical composition and antioxidant activity of the essential oil of endemic Viola tianshanica.
Natural product research (2011-10-21)
Jin Yang, Zhen Qu, Yu-Ling Xiao, Guo-Fu Qiu, Tao Zhang, Zhong-Yuan Wu, Xian-Ran He, Xian-Ming Hu
ABSTRAKT
The composition and in vitro antioxidant activities of the essential oil and methanol extract of the aerial parts of Viola tianshanica were evaluated in this research. GC-MS analysis of the essential oil resulted in the identification of 15 constituents, representing 89.67% of the oil. The major compounds detected in the essential oil were dibutyl phthalate (15.19%), hexadecanoate methyl (8.65%), n-hexadecanoic acid (3.07%) and 2,3-pentanedione (2.62%). Essential oil and methanol extract were tested for their antioxidant activities using 1,1-diphenyl-2-picryl-hydrazyl free radical scavenging and β-carotene linoleic acid assay. In addition, the total phenol of essential oil, polar subfraction and non-polar subfraction were determined.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,3-Pentanedione, natural, ≥96%, FG
Sigma-Aldrich
2,3-Pentanedione, 97%
Sigma-Aldrich
2,3-Pentanedione, ≥96%, FCC, FG