Merck
Solid/Liquid phase diagram of the ammonium sulfate/maleic acid/water system.
The journal of physical chemistry. A (2011-10-25)
Keith D Beyer, Jason R Schroeder, Christian S Pearson
ABSTRAKT
We have studied the low temperature phase diagram and water activities of the ammonium sulfate/maleic acid/water system using differential scanning calorimetry and infrared spectroscopy of thin films. Using the results from our experiments, we have mapped the solid/liquid ternary phase diagram, determined the water activities based on the freezing point depression, and determined the ice/maleic acid phase boundary as well as the ternary eutectic composition and temperature. We also compare our results to the predictions of the extended AIM aerosol thermodynamics model and find good agreement for the ice melting points in the ice primary phase field of this system; however significant differences were found with respect to phase boundaries, maleic acid dissolution, and ammonium sulfate dissolution.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Maleic acid sodium salt, ~99%
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium maleate dibasic, purum p.a., anhydrous, ≥98.0% (NT)
Supelco
Maleic acid, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland