Merck
Discovery of α,β- and α,γ-diamino acid scaffolds for the inhibition of M1 family aminopeptidases.
ChemMedChem (2011-10-26)
Rajesh Gumpena, Chandan Kishor, Roopa Jones Ganji, Anthony Addlagatta
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-2,4-Diaminobutyric acid dihydrochloride, ≥95.0%