Merck
Nanostructuration strategies to enhance microelectrode array (MEA) performance for neuronal recording and stimulation.
Journal of physiology, Paris (2011-10-27)
Matthias Heim, Blaise Yvert, Alexander Kuhn
ABSTRAKT
Microelectrode arrays (MEAs) are widely used tools for recording and stimulating extracellular neuronal activity. Major limitations when decreasing electrode size in dense arrays are increased noise level and low charge injection capability. Nanostructuration of the electrode sites on MEAs presents an efficient way to overcome these problems by decreasing the impedance of the electrode/solution interface. Here, we review different techniques used to achieve this goal including template assisted electrodeposition for generating macro- and mesoporous films, immobilization of carbon nanotubes (CNTs) and deposition of conducting polymers onto microelectrodes. When tested during in vitro and in vivo measurements, nanostructured MEAs display improved sensitivity during recording of neuronal activity together with a higher efficiency in the stimulation process compared to conventional microelectrodes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iridium(IV) oxide, 99.9% trace metals basis