Merck
Genome sequence of Bacillus pumilus S-1, an efficient isoeugenol-utilizing producer for natural vanillin.
Journal of bacteriology (2011-11-01)
Fei Su, Dongliang Hua, Zhaobin Zhang, Xiaoyu Wang, Hongzhi Tang, Fei Tao, Cui Tai, Qiulin Wu, Geng Wu, Ping Xu
ABSTRAKT
Bacillus pumilus S-1 is an efficient isoeugenol-utilizing producer of natural vanillin. The genome of B. pumilus S-1 contains the epoxide hydrolase and six candidate monooxygenases that make it possible to explore the mechanism involved in conversion of isoenguenol to vanillin in the B. pumilus strain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Isoeugenol, analytical standard
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG