Merck
Septic arthritis due to Fusobacterium nucleatum in an immunocompetent patient.
Reumatologia clinica (2011-11-18)
Carmen Carrasco Cubero, Priscila Zamora Red, José Javier Salaberri Maestrojuan, M Dolores López Prieto
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cloxacillin sodium salt monohydrate, β-lactamase-resistant antibiotic
Supelco
Cloxacillin sodium salt monohydrate, VETRANAL®, analytical standard