Merck
Ethylmalonic acid impairs brain mitochondrial succinate and malate transport.
Molecular genetics and metabolism (2011-12-03)
Alexandre Umpierrez Amaral, Cristiane Cecatto, Estela Natasha Brandt Busanello, César Augusto João Ribeiro, Daniela Rodrigues Melo, Guilhian Leipnitz, Roger Frigério Castilho, Moacir Wajner
ABSTRAKT
Tissue accumulation and high urinary excretion of ethylmalonic acid (EMA) occur in ethylmalonic encephalopathy (EE) and short chain acyl-CoA dehydrogenase deficiency (SCADD). Although these autosomal recessive disorders are clinically characterized by neurological abnormalities, the mechanisms underlying the brain damage are poorly known. Considering that little is known about the neurotoxicity of EMA and that hyperlacticacidemia occurs in EE and SCADD, we evaluated the effects of this metabolite on important parameters of oxidative metabolism in isolated rat brain mitochondria. EMA inhibited either ADP-stimulated or uncoupled mitochondrial respiration supported by succinate and malate, but not by glutamate plus malate. In addition, EMA mildly stimulated oxygen consumption by succinate-respiring mitochondria in resting state. Methylmalonic acid (MMA), malonic acid (MA) and butylmalonic acid (BtMA) had a similar effect on ADP-stimulated or uncoupled respiration. Furthermore, EMA-, MMA- and BtMA-induced inhibitory effects on succinate oxidation were significantly minimized by nonselective permeabilization of the mitochondrial membranes by alamethicin, whereas MA inhibitory effect was not altered. In addition, MA was the only tested compound that reduced succinate dehydrogenase activity. We also observed that EMA markedly inhibited succinate and malate transport through the mitochondrial dicarboxylate carrier. Mitochondrial membrane potential was also reduced by EMA and MA, but not by MMA, using succinate as electron donor, whereas none of these compounds was able to alter the membrane potential using glutamate plus malate as electron donors. Taken together, our results strongly indicate that EMA impairs succinate and malate uptake through the mitochondrial dicarboxylate carrier.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylmalonic acid, 97%