Merck
Sternoclavicular septic arthritis and empyema.
Reumatologia clinica (2011-12-14)
María José Moreno Martínez, Manuel José Moreno Ramos, Luis F Linares Ferrando, Carlos Marras Fernandez-Cid, Manuel Castaño Sanchez, Elena Peñas Martínez
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cloxacillin sodium salt monohydrate, β-lactamase-resistant antibiotic
Supelco
Cloxacillin sodium salt monohydrate, VETRANAL®, analytical standard