Merck
Antimicrobial potential of 3-hydroxy-2-methylene-3-phenylpropionic acid derivatives.
Acta pharmaceutica (Zagreb, Croatia) (2011-12-29)
Soni A Singh, Sujata V Bhat
ABSTRAKT
Twenty Baylis-Hillman adducts were synthesized from different aromatic aldehydes and activated vinyl derivatives. The adducts, which are differently substituted 3-hydroxy-2-methylene-3-phenylpropionic acid derivatives, were screened for their antimicrobial activity in vitro by the serial dilution method. Many of these molecules displayed potent antibacterial and antifungal activities. The ease of synthesis from low-cost starting materials along with potent antimicrobial activity of these molecules provide the lead for further improvement of activity and reflect the possibility of therapeutic use.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrocinnamic acid, 99%
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG