- Improvement of modified charcoal-cefoperazone-deoxycholate agar by supplementation with a high concentration of polymyxin B for detection of Campylobacter jejuni and C. coli in chicken carcass rinses.
Improvement of modified charcoal-cefoperazone-deoxycholate agar by supplementation with a high concentration of polymyxin B for detection of Campylobacter jejuni and C. coli in chicken carcass rinses.
Applied and environmental microbiology (2012-01-03)
Jung-Whan Chon, Ji-Yeon Hyeon, Jin-Hyuk Yim, Jong-Hyun Kim, Kwang-Young Song, Kun-Ho Seo
PMID22210208
ABSTRAKT
Modified charcoal-cefoperazone-deoxycholate agar (mCCDA) was improved by supplementation with a high concentration of polymyxin B. The ability of the supplemented medium to isolate Campylobacter jejuni and C. coli from chicken carcass rinses was compared to that of Campy-Cefex agar and mCCDA. Modification of mCCDA with increased polymyxin B yielded a significantly (P < 0.05) higher isolation rate and greater selectivity than those achieved using Campy-Cefex agar and mCCDA.