Merck
Narrow-band ultraviolet B treatment for diphenylcyclopropenone-induced vitiliginous lesions.
Acta dermato-venereologica (2012-01-05)
Yu-Jin Oh, Min-Kyung Shin, Mu-Hyoung Lee
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diphenylcyclopropenone, 98%