Merck
Self-assembled pullulan-silica oxygen barrier hybrid coatings for food packaging applications.
Journal of agricultural and food chemistry (2012-01-06)
Stefano Farris, Laura Introzzi, José Maria Fuentes-Alventosa, Nadia Santo, Roberto Rocca, Luciano Piergiovanni
ABSTRAKT
The scope of this study encompassed the evaluation of pullulan as a suitable biopolymer for the development of oxygen barrier coatings to be applied on poly(ethylene terephthalate) (PET), especially for food packaging applications. To enhance the oxygen barrier properties of the organic phase (pullulan) even at high relative humidity values, an inorganic phase (silica), obtained through in situ polymerization, was also utilized to obtain hybrid coatings via the sol-gel technique. Transmission electron microscopy (TEM) images and Fourier transform infrared (FT-IR) spectra showed that mixing the two phases yielded a three-dimensional hybrid network formed by self-assembly and mediated by the occurrence of new hydrogen-bond interactions at the intermolecular level, although the formation of new covalent bonds could not be excluded. The deposition of the hybrid coatings decreased the oxygen transmission rate (OTR) of the plastic substrate by up to 2 orders of magnitude under dry conditions. The best performance throughout the scanned humidity range (0%-80% relative humidity) was obtained for the formulation with the lowest amount of silica (that is, an organic/inorganic ratio equal to 3).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Pullulan Standard Set Mp ~350 - 700′000, set of analytical standards, suitable for gel permeation chromatography (GPC)
Sigma-Aldrich
Pullulan, suitable for substrate for pullulanase