Merck
Photosensitization mechanisms of triplet excited state beta-lapachone. A density functional theory study.
Natural product communications (2012-01-10)
Liang Shen
ABSTRAKT
Beta-Lapachone is a natural product with multiple pharmacological activities and mechanistic studies indicated that reactive oxygen species (ROS) generated by beta-lapachone play significant roles in its pharmacological actions. As photosensitization is an important ROS-generating pathway, in the present work, the photosensitization mechanisms of beta-lapachone are explored on the basis of density functional theory estimated triplet excited state characters. Starting from triplet excited state beta-lapachone, the possible generating pathways of 1O2 and O2*- are elucidated and the solvent effects on the photosensitizing reactions are also discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Lapachone, ≥98% (TLC)