Merck
  • FeCl3⋅6H2O-catalyzed intermolecular-cascade cyclization of acetoacetanilide: aldehyde-tuned synthesis to valuable 2-pyridone analogues.
FeCl3⋅6H2O-catalyzed intermolecular-cascade cyclization of acetoacetanilide: aldehyde-tuned synthesis to valuable 2-pyridone analogues.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-01-24)
Tista Sengupta, Krishnanka S Gayen, Palash Pandit, Dilip K Maiti
ABSTRAKT
The first ever breakthrough toward activation of β-ketoacetanilide and subsequent C-C and C-N bond-forming intermolecular-cascade cyclization processes is demonstrated by development of the unprecedented Lewis acid property of non-toxic FeCl(3)⋅6H(2)O. Aromatic, aliphatic, α,β-unsaturated, chiral sugar-based and chromone aldehydes were regio- and stereoselectively cyclized with acetoacetanilides toward construction of valuable N-containing highly functionalized 2-pyridones (see scheme for an example).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Hydroxypyridine, 97%
Sigma-Aldrich
Acetoacetanilide, ≥99.5%
Levetiracetam impurity C, European Pharmacopoeia (EP) Reference Standard