Merck
Prevalence of desloratadine poor metabolizer phenotype in healthy Jordanian males.
Biopharmaceutics & drug disposition (2012-01-25)
Nancy Hakooz, Isam I Salem
ABSTRAKT
To study the prevalence of desloratadine slow metabolizer phenotype among a group of healthy Jordanian male volunteers. A total of 62 healthy Jordanian male volunteers were included in this study. A single 5 mg desloratadine oral tablet was given and blood samples were taken to determine the desloratadine and 3-hydroxydesloratadine (3-OH-desloratadine) concentrations using a specific liquid chromatography-mass spectrometric method (LC/MS/MS). The determination of pharmacokinetic parameters of all the individuals was determined by using Kinetica® program version 4.1. Poor metabolizers or slow metabolizers of desloratadine were determined as individuals having a 3-OH-desloratadine to desloratadine exposure ratio lower than 10% or a desloratadine half-life ≥ 50 h. Among the 62 volunteers who participated in the study there were only two volunteers who were labeled as desloratadine slow metabolizers, giving a prevalence of 3.2%. The maximum plasma concentrations (C(max)) were similar in the extensive and slow metabolizers groups but a longer time (t(max)) was needed to achieve this concentration in one of the volunteers who was a desloratadine slow metabolizer. The incidence of the poor metabolizer phenotype of desloratadine in the Jordanian population studied is similar to certain ethnic groups (e.g. Asian, Caucasians and Hispanic); however, it is lower than other populations (e.g. American Indians and Black).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Loratadine for system suitability, European Pharmacopoeia (EP) Reference Standard
Loratadine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Loratadine, ≥98% (HPLC), powder