Merck
Ketonization of enols in aqueous solution: is carbon protonation always rate-determining?
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2012-01-27)
Anna Paola Pelliccioli, Peter Šebej, Jakob Wirz
ABSTRAKT
The pH-rate profiles for the ketonization of the (E)- and (Z)-photoenols of o-methylacetophenone (MA) in aqueous solution were determined by nanosecond laser flash photolysis. Carbon protonation of the enol anions of MA by solvent water is exceptionally fast, k(0)'(K)≈ 2.0 × 10(7) s(-1), too fast to permit establishment of the acid-base equilibrium on the enol oxygen prior to ketonization. Analysis of the pH-rate profile of the (E)-enol using the common assumption of rate-determining carbon protonation would lead to an erroneous value for the acidity constant of that enol, pK(a,c)(E) = 11.3, which is too high by about two pK units.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetophenone, ≥98%, FG