Merck
Preclinical evaluation of the antineoplastic efficacy of 7-(2-hydroxyethyl)theophylline on melanoma cancer cells.
Melanoma research (2012-02-02)
Alessandro Lentini, Claudio Tabolacci, Alessandra Nardi, Palma Mattioli, Bruno Provenzano, Simone Beninati
ABSTRAKT
Differentiation-based therapeutics are an underutilized but potentially a significant option for cancer treatment. The effect of methylxanthines on melanoma cell differentiation has been well documented. We report the in-vitro and in-vivo anticancer potential of a theophylline analogue, 7-(2-hydroxyethyl)theophylline (HET), on murine B16-F10 and human Sk-Mel 110 metastatic melanoma cell lines. The effects on cell proliferation were related to the induction of differentiation, demonstrated as increased intracellular transglutaminase activity. The involvement of this methylxanthine in the control of the in-vitro adhesion and in the in-vivo metastastic spread of melanoma cells was further investigated. HET oral administration of C57BL6/N mice intravenously injected with B16-F10 cells markedly reduced lung metastases frequency. The overall results demonstrated that HET possesses a remarkable in-vivo antimetastatic capability.