Merck
Increased [³H]D-aspartate release and changes in glutamate receptor expression in the hippocampus of the mnd mouse.
Journal of neuroscience research (2012-02-04)
Paolo Bigini, Marco Milanese, Fabrizio Gardoni, Annalisa Longhi, Tiziana Bonifacino, Sara Barbera, Elena Fumagalli, Monica Di Luca, Tiziana Mennini, Giambattista Bonanno
ABSTRAKT
Neuronal ceroid lipofuscinoses (NCLs) are a group of hereditary childhood diseases characterized mainly by lipopigment accumulation and a multisystemic pattern of symptoms including mental retardation, seizures, motor impairment, and blindness. The mnd mouse, carrying a mutation in the Cln8 gene, has been proposed as a model of epilepsy with mental retardation (EPMR, ornorthern epilepsy). We recently showed neuronal hyperexcitability and seizure hypersusceptibility in mnd mice. To elucidate the cellular mechanisms related to hippocampal hyperexcitability, the glutamatergic transmission and the expression of postsynaptic glutamate receptors were investigated in hippocampus. A significant increase in either spontaneous or KCl-stimulated overflow of [³H]D-aspartate was found in mnd mice compared with controls. This increase was maintained after DL-threo-β-benzyloxyaspartic acid (TBOA) treatment, suggesting a nonrelevant role for transporter-mediated release and supporting the involvement of exocytotic [³H]D-aspartate release. Accordingly, Ca²⁺-dependent overflow induced by ionomycin was also increased in mnd mice. Levels of glutamate 1-3 AMPA receptor subunits were increased, and levels of the NR2A NMDA receptor subunit were decreased in the hippocampus of mnd mice, suggesting an adaptive response to glutamate overstimulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Aspartic acid, ReagentPlus®, 99%
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Glutamate Receptor 4 Antibody, Chemicon®, from rabbit