Merck
Experimental results using 3-bromopyruvate in mesothelioma: in vitro and in vivo studies.
Journal of bioenergetics and biomembranes (2012-02-11)
Philippe Icard, Icard Philippe, Xiao-Dong Zhang, Zhang Xiao-Dong, Edwige Lemoisson, Lemoisson Edwige, Marie-Hélène Louis, Louis Marie-Hélène, Stéphane Allouche, Allouche Stéphane, Hubert Lincet, Lincet Hubert, Laurent Poulain, Poulain Laurent
ABSTRAKT
Over many years we have taken advantage of the special metabolism of cancer cells involving an increased consumption of glucose associated with lactic acid production even in the presence of oxygen, a phenomenon referred to as the "Warburg effect", to counteract cancer cell growth. We have tested 3-bromopyruvate (3-BrPA), an inhibitor of pyruvate-associated reactions. Firstly, we tested this agent, in vitro, in two mesothelioma cell lines. Cellular response would appear to depend on the mode of administration (immediately or 24 h after seeding). Depending on the line, 3-BrPA induced a cytostatic or cytotoxic effect. This effect was accompanied by cell death induction even in cells highly refractory to cisplatin. Mitochondrial apoptotic death appeared to involve both lines; however, a different death pathway such as necrosis cannot be excluded. Interestingly, 3-BrPA leads to a diminution of the expression of the anti-apotptoic protein Mcl-1. We then tested 3-BrPA in vivo. Survival of nude mice bearing human mesothelioma was significantly prolonged (p < 0.0001). Toxicity and clinical studies should be performed to test 3- BrPA as local therapy for patients suffering from pleural or peritoneal mesothelioma. Association with cisplatin should be particularly considered.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bromopyruvic acid, ≥98.0%