Merck
  • In vitro degradation and cytocompatibility of dicalcium phosphate dihydrate cements prepared using the monocalcium phosphate monohydrate/hydroxyapatite system reveals rapid conversion to HA as a key mechanism.
In vitro degradation and cytocompatibility of dicalcium phosphate dihydrate cements prepared using the monocalcium phosphate monohydrate/hydroxyapatite system reveals rapid conversion to HA as a key mechanism.
Journal of biomedical materials research. Part B, Applied biomaterials (2012-02-11)
Daniel L Alge, W Scott Goebel, Tien-Min Gabriel Chu
ABSTRAKT
We previously showed that dicalcium phosphate dihydrate (DCPD) cements can be prepared using monocalcium phosphate monohydrate (MCPM) and hydroxyapatite (HA). In this study, we have characterized the degradation properties and biocompatibility of these novel cements. To study the degradation properties, cements were prepared using MCPM:HA molar ratios of 4:1, 2:1, 2:3, and 2:5. Degradation was evaluated in vitro by static soaking in PBS, and changes in pH, mass, compressive strength, and composition were monitored. Conversion of DCPD to HA was noted in the 4:1 group, which initially consisted of pure DCPD. However, the 2:1 group, which initially consisted of DCPD and an intermediate amount of unreacted HA, underwent rapid conversion to HA associated with significantly greater pH drop and mass loss as well as a complete loss of mechanical integrity. On the basis of these results, we directly compared the cytocompatibility of 2:1 MCPM:HA cements to DCPD cements prepared with an equivalent percent molar excess of β-tricalcium phosphate (β-TCP) using an in vitro cell viability assay. Viability of cells co-cultured with 2:1 MCPM:HA cements was significantly reduced after just 48 h, while viability of cells cultured with the β-TCP-based cements was no different from control cells. In conclusion, this study demonstrates that conversion to HA plays an important role in the degradation of DCPD cements prepared with the MCPM/HA system, affecting both physical properties and cytocompatibility. These results could have important clinical implications for MCPM/HA cements.