Merck
Synthesis of 2,3,5,6-tetrasubstituted tetrahydropyrans via (3,5)-oxonium-ene reaction.
Organic & biomolecular chemistry (2012-02-22)
Pipas Saha, Anup Bhunia, Anil K Saikia
ABSTRAKT
An efficient method for the synthesis of 2,3,5,6-tetrasubstituted tetrahydropyrans has been developed from the reaction of aldehydes and ethyl 2-(1-hydroxyalkyl/hydroxy(phenyl)methyl)-5-methylhex-4-enoate using (3,5)-oxonium-ene reaction promoted by boron trifluoride etherate in good yields under mild conditions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%