Merck
[Influence of natural complex compounds with hipolipidemic activity on expression of bacteria virulence factors].
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic] (2012-03-01)
V Iu Gotovtseva, N É Grammatikova, M V Bibikova, A V Katlinskiĭ
ABSTRAKT
Natural complex compounds with hipolipidemic activity, having considerable inhibitory effect on expression of bacteria virulence factors were isolated. Inhibitory properties of the compounds with respect to pyocyanine and protease formation, as well as their influence on the quorum sensing mechanism in Chromobacterium violacium were shown.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyocyanin, Ready Made Solution from Pseudomonas aeruginosa, 5 mg/mL in DMSO
Sigma-Aldrich
Pyocyanin, from Pseudomonas aeruginosa, ≥98% (HPLC)