Merck
Looking for the physiological role of anthocyanins in the leaves of Coffea arabica.
Photochemistry and photobiology (2012-03-01)
Adilson Pereira Domingues Júnior, Milton Massao Shimizu, Jullyana Cristina Magalhães Silva Moura, Rodrigo Ramos Catharino, Rômulo Augusto Ramos, Rafael Vasconcelos Ribeiro, Paulo Mazzafera
ABSTRAKT
The aim of this study was to determine which anthocyanins are related to the purple coloration of young leaves in Coffea arabica var. Purpurascens and assess their impact on photosynthesis as compared to C. arabica var. Catuaí, with green leaves. Two delphinidin glicosides were identified and histological cross-sections showed they were located throughout the adaxial epidermis in young leaves, disappearing as the leaves mature. Regardless the irradiance level, the photosynthetic performance of Purpurascens leaves did not differ from that observed in leaves of the Catuaí variety, providing no evidence that anthocyanins improve photosynthetic performance in coffee plants. To analyze the photoprotective action of anthocyanins, we evaluated the isomerization process for chlorogenic acids (CGAs) in coffee leaves exposed to UV-B radiation. No differences were observed in the total concentration of phenolic compounds in either variety before or after the UV treatment; however, we observed less degradation of CGA isomers in the Purpurascens leaves and a relative increase of cis-5-caffeoylquinic acid, a positional isomer of one of the most abundant form of CQA in coffee leaves, trans-5-caffeoylquinic acid, suggesting a possible protective role for anthocyanins in this purple coffee variety.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Delphinidin chloride, analytical standard