Merck
Effects of parabens on apoptosis induced by serum-free medium.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2012-03-02)
Mari Egawa, Kentaro Aoki, Yongkun Sun, Toshiyuki Hosokawa, Takeshi Saito, Masaaki Kurasaki
ABSTRAKT
Alkyl esters of p-hydroxybenzoic acids (parabens), an endocrine disrupter, are used as preservatives in cosmetics and foods. In this study, to understand the relationship between parabens and differentiation in infants, the effects of parabens on apoptosis induced by serum deprivation in PC12 cells were investigated. In addition, apoptosis-related factors were assayed. As results, a tendency toward enhancement of apoptosis was observed in the cells cultured in the serum-free medium with methylparaben, and this tendency was suggested to be related to the contents of BAD, a pro-apoptotic protein. Butylparaben did not show any tendency to enhance apoptosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)