Merck
  • Morin attenuates blood pressure and oxidative stress in deoxycorticosterone acetate-salt hypertensive rats: a biochemical and histopathological evaluation.
Morin attenuates blood pressure and oxidative stress in deoxycorticosterone acetate-salt hypertensive rats: a biochemical and histopathological evaluation.
Metabolism: clinical and experimental (2012-03-06)
Pichavaram Prahalathan, Subramanian Kumar, Boobalan Raja
ABSTRAKT
The present study was designed to evaluate the antihypertensive and antioxidant effect of morin, a flavonoid against deoxycorticosterone acetate (DOCA)-salt induced hypertension in male Wistar rats. Hypertension was induced in uninephrectomized rats (UNX) by weekly twice subcutaneous injection of DOCA (25mg/kg) and 1% NaCl in the drinking water for six consecutive weeks. The DOCA-salt hypertensive rats showed significant (P < .05) increase in the systolic and diastolic blood pressure, heart rate, water intake and organ weights (kidney, heart, aorta and liver). DOCA-salt hypertensive rats also showed significant (P < .05) increase in the levels of thiobarbituric acid reactive substances, lipid hydroperoxides and conjugated dienes in plasma and tissues (kidney, heart, aorta and liver), and significant (P < .05) decrease in the body weight, nitrite and nitrate levels in plasma and heart. Furthermore, the activities of enzymic antioxidants such as superoxide dismutase, catalase and glutathione peroxidase in erythrocyte and tissues and the levels of non-enzymic antioxidants such as reduced glutathione, vitamin C and vitamin E in plasma and tissues were significantly (P < .05) decreased in DOCA-salt rats. Morin supplementation (50mg/kg) daily for six weeks brought back all the above parameters to near normal level. The above findings were confirmed by the histopathological examination. No significant (P < .05) effect was observed in UNX-rats treated with morin (50mg/kg). These results suggest that morin acts as an antihypertensive and antioxidant agent against DOCA-salt induced hypertension.