Merck
Myocardial imaging for mitochondrial membrane potential.
JACC. Cardiovascular imaging (2012-03-17)
H William Strauss, Heiko Schöder
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tetraphenylphosphonium bromide, 97%
Sigma-Aldrich
Tetraphenylphosphonium chloride, 98%