Merck
One-pot synthesis of 4-substituted isoquinolinium zwitterionic salts by metal-free C-H bond activation.
Chemical communications (Cambridge, England) (2012-03-30)
Hong Hou, Yuan Zhang, Chao-Guo Yan
ABSTRAKT
The unprecedented isoquinolinium zwitterionic salts with an unusual C-4 substitution pattern were efficiently prepared via the multicomponent reaction of in situ formed N-benzylisoquinolinium bromide with aromatic aldehydes and cyclic 1,3-dicarbonyl compounds.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isoquinoline, 97%