Merck
ALSUntangled 15: coconut Oil.
Amyotrophic lateral sclerosis : official publication of the World Federation of Neurology Research Group on Motor Neuron Diseases (2012-04-05)
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Coconut oil, virgin, Certified organic (NOP)
Sigma-Aldrich
Coconut oil from Cocos nucifera, low-melting solid