Merck
Biodegradation of 2-chlorophenol (2CP) in an anaerobic sequencing batch reactor (ASBR).
Water science and technology : a journal of the International Association on Water Pollution Research (2012-05-02)
Lizeth Beristain-Montiel, Jorge Gómez, Oscar Monroy, Flor de Maria Cuervo-López, Florina Ramírez-Vives
ABSTRAKT
The aim of this study is to contribute to the knowledge about anaerobic digestion of 2-chlorophenol (2CP) in an anaerobic sequencing batch reactor (ASBR). Two reactors were set up (ASBR(A) and ASBR(B)). The ASBR(A) was fed with 2-chlorophenol (28-196 mg 2CP-C/L) and no other exogenous electron donor. The ASBR(B) was fed with a mixture of 2CP (28-196 mg 2CP-C/L) and phenol (28-196 mg phenol-C/L) as an electron donor. The process evaluation was conducted by three means: first by substrate consumption efficiency (E(2CP)), second, by biogas yield (Y(biogas-C/2CP-C)) and third, by the specific consumption rates (q(2CP)) as response variables. The 2CP consumption efficiency (90 ± 0.4%) was not influenced by the increase in the concentrations tested. In both reactors ASBR(A) and ASBR(B), both concentration as well as speed increased. Concentration increased from 28 to 114 mg 2CP-C/L. The specific consumption rate (q(2CP)) values were fivefold higher. However, a decrease of 37% was observed at 140 mg 2CP-C/L and one of 72% at 196 mg 2CP-C/L. The biogas yields (0.80 ± 0.06) remained stable in both reactors. In both reactors the biogas yield decreased to 78 ± 3% at 196 mg 2CP-C/L. We might assume this decrease was due to the accumulation of VFA. Finally, poor sludge settleability was determined only in the SBR(B) reactor at 140 and 196 mg 2CP-C/L. An increase was observed in both SVI ≤ 140 ± 5 mL/g and over exopolymeric protein ≤120 mg EP/L.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Chlorophenol, ≥99%
Sigma-Aldrich
2-Chlorophenol, 98%