- Printable superhydrophilic-superhydrophobic micropatterns based on supported lipid layers.
Printable superhydrophilic-superhydrophobic micropatterns based on supported lipid layers.
Langmuir : the ACS journal of surfaces and colloids (2012-05-19)
Junsheng S Li, Erica Ueda, Asritha Nallapaneni, Linxian X Li, Pavel A Levkin
PMID22594681
ABSTRAKT
A new and simple method for creating superhydrophilic micropatterns on a superhydrophobic surface is demonstrated. The method is based on printing an "ink", an ethanol solution of a phospholipid, onto a porous superhydrophobic surface and, thus, is compatible with a variety of commonly available printing techniques.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor