Merck
  • Trace determination of dichlorvos in environmental samples by room temperature ionic liquid-based dispersive liquid-phase microextraction combined with HPLC.
Trace determination of dichlorvos in environmental samples by room temperature ionic liquid-based dispersive liquid-phase microextraction combined with HPLC.
Journal of chromatographic science (2012-05-24)
Songhui Wang, Bingren Xiang, Qianqian Tang
ABSTRAKT
Using 1-butyl-3-methylimidazolium hexafluorophosphate ([BMIM][PF6]) room temperature ionic liquid (RTIL) as extraction solvent, tetrahydrofuran (THF) as disperser solvent, the organophosphorus pesticide dichlorvos in water was determined by dispersive liquid-liquid microextraction (DLLME) combined with high-performance liquid chromatography. Factors affecting RTIL-DLLME (type of disperser solvent, amount of RTIL, volume of disperser solvent, percentage of NaCl and volume and pH of water sample) were optimized by the single-factor method, obtaining the most favorable results when using 65 µL of [BMIM][PF6] and 260 µL of THF to extract the compound from an 8-mL water sample at pH 5.0 containing 25% (w/v) of NaCl. Under these optimum conditions, an enrichment factor of 215-fold was obtained. The calibration curves were linear in the concentration range of 2-1,000 µg/L. The limit of detection calculated at a signal-to-noise ratio of 3 was 0.2 µg/L. The relative standard deviations (RSD) for six replicate experiments at 20, 100 and 200 µg/L concentration levels were 1.8%, 1.3% and 1.3 %, respectively. Then the proposed method was applied to the analysis of three different water sample sources (tap, farm and rain water) and the relative recoveries and RSD of spiked water samples were 95.6-102.4% and 0.6-3.1%, respectively, at three different concentration levels of 20, 100 and 200 µg/L.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Dichlorvos, PESTANAL®, analytical standard