Merck
[The medical treatment of Kim Phúc at the BG Unfallklinik Ludwigshafen].
Handchirurgie, Mikrochirurgie, plastische Chirurgie : Organ der Deutschsprachigen Arbeitsgemeinschaft fur Handchirurgie : Organ der Deutschsprachigen Arbeitsgemeinschaft fur Mikrochirurgie der Peripheren Nerven und Gefasse : Organ der V... (2012-05-24)
J Kiefer, A Daigeler, M Lehnhardt
ABSTRAKT
The Vietnam War was a military conflict in Vietnam during the Cold War that followed the First Indochina War. This war was fought between North Vietnam, supported by its communist allies, and the government of South Vietnam, supported by the USA and other anti-communist countries. Kim Phúc is the child depicted in the Pulitzer Prize winning photograph taken on June 8, 1972 by AP photographer Nick Út. The iconic photo shows her at about nine years of age running naked on a road amid the chaos after being severely burned by a napalm attack. After 14 months of hospital stay and 17 surgical procedures Kim Phúc was able to return home. Since then, she was used as a propaganda symbol by the communist government of Vietnam. To continue her studies, Kim was granted permission to move to Cuba where she met her future husband. However, the sequelae of her burn wounds affected her everyday life enormously. In 1984, with the support of the international aid organization "terre des hommes" and the German magazine "STERN", Kim Phúc got the opportunity to meet and get treated by Professor Zellner. Professor Peter Rudolph Zellner was the first chief of the Department of Hand, Plastic and Reconstructive Surgery, Burn Center, BG Trauma Center Ludwigshafen, and one of the founder members of the German Society of Plastic Surgeons. The reconstructive surgeries provide Kim Phúc an almost normal life. Later on, she was involved in international aid organizations; she was named a UNESCO Goodwill Ambassador and she was awarded several honorary Doctorates of Law. Kim Phúc became a Canadian citizen. Today, she lives with her husband and two children in Ontario, Canada.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Naphthenic acid, technical