Merck
The potential clinical impact of quantum dots.
Nanomedicine (London, England) (2012-05-29)
Swadeshmukul Santra
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trioctylphosphine oxide, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%
Sigma-Aldrich
Trioctylphosphine oxide, for extraction analysis, ≥98.5%