Merck
Polytherapy for migraine prophylaxis.
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology (2012-06-01)
G Casucci, V Villani, D Cologno, F D'Onofrio
ABSTRAKT
Migraine is a chronic neurological disorder with episodic manifestations, progressive in some individuals. Preventive treatment is recommended for patients with frequent or disabling attacks. A sizeable proportion of migraineurs in need of preventive treatment does not significantly benefit from monotherapy. This short review evaluates the role of pharmacological polytherapy in migraine prevention.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nortriptyline hydrochloride, ≥98% (HPLC), powder
Nortriptyline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Nortriptyline hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®