Merck
Reaction-based two-photon probes for in vitro analysis and cellular imaging of monoamine oxidase activity.
Chemical communications (Cambridge, England) (2012-06-02)
Dokyoung Kim, Sunderraman Sambasivan, Hyoseok Nam, Ki Hean Kim, Jin Yong Kim, Taiha Joo, Kyung-Ha Lee, Kyong-Tai Kim, Kyo Han Ahn
ABSTRAKT
Reaction-based fluorescent probes for monoamine oxidases A and B are developed based on a new two-photon absorbing compound and its precursor. The probes show turn-on fluorescence response to the enzymes owing to the two-photon absorbing compound produced by the enzymatic activity, as monitored by one- as well as two-photon microscopy for the first time.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acrylonitrile solution, certified reference material, 5000 μg/mL in methanol
Supelco
Acrylonitrile, analytical standard
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor