Merck
[Simultaneous determination of isoorientin, scutellarin and cynaroside in rat plasma and pharmacokinetics by UPLC-MS/MS].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-06-07)
Yong Huang, Feng He, Lin Zheng, Zhirong Zhang, Yanyu Lan, Yonglin Wang
ABSTRAKT
To establish a UPLC-MS/MS analysical method for simultaneous determination of concentrations of isoorientin, scutellarin and cynaroside in rat plasma and to study their pharmacokinetic characteristics after intravenous injection of 3 doses of Fufang Hongcao in rats. Acidified plasma samples were precipitated for protein with methanol. Waters Acquity BEH C18 column was adopted for spectrum, with mobile phase as 0. 1% formic acid acetonitrile-0. 1% formic acid-water gradient elution. Detection was carried out by the multiple reaction monitoring (MRM) positive ion mode with ESI ionization source. Three flavonoids show a good linear relationship, with the extraction recovery ranging between 78.56% and 101.91% and a high intra-and inter-day precisions and accuracy. The MRT of the three flavonoids were all lower than 22 min in rats. The above men tioned method is so specific, rapid, sensitive that it is suitable for pharmacokinetic studies of Fufang Hongcao injection in rats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Luteolin 7-glucoside, analytical standard
Sigma-Aldrich
Luteolin 7-O-β-D-glucoside, ≥98.0% (HPLC)
7-glukozyd luteoliny, primary reference standard
Sigma-Aldrich
Isoorientin, ≥98% (HPLC)
Supelco
Scutellarin, analytical standard
Isoorientin, primary reference standard