Merck
Evaluation of extended release brimonidine intravitreal device in normotensive rabbit eyes.
Acta ophthalmologica (2012-06-09)
Sunil P Deokule, Judit Z Baffi, Hong Guo, Marty Nazzaro, Hiroki Kaneko
ABSTRAKT
To evaluate the safety profile of a brimonidine extended release intravitreal implant, in normotensive rabbit eyes. Devices were made from hollow poly-l-lactic acid (PLA) tubes and contained hundred micrograms of brimonidine pamoate. Device was injected intravitreally in one eye of 12 New Zealand pigmented rabbits, whereas other eye was injected with a sham implant in masked fashion. Ocular examination was conducted at baseline and months 1, 3 and 6 including dilated fundus examination and electro-retinogram (ERG). Four rabbits were sacrificed at each time-point for retinal histology. ERG data were compared between groups and time-points using anova. No complications were reported from either eye of any rabbits over a 6-month period. Photopic A wave was reduced in the control eye at 1 month compared with baseline (p < 0.01). There was no significant difference in other ERG parameters between the groups at different time-points. Gross retinal histology was normal at all time-points. Extended release intravitreal brimonidine device was found to be safe and in normotensive rabbit eyes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
UK 14,304