Merck
Haloenol pyranones and morpholinones as antineoplastic agents of prostate cancer.
Bioorganic & medicinal chemistry letters (2012-06-09)
Jason N Mock, John P Taliaferro, Xiao Lu, Sravan Kumar Patel, Brian S Cummings, Timothy E Long
ABSTRAKT
Haloenol pyran-2-ones and morpholin-2-ones were synthesized and evaluated as inhibitors of cell growth in two different prostate human cancer cell lines (PC-3 and LNCaP). Analogs derived from L- and D-phenylglycine were found to be the most effective antagonists of LNCaP and PC-3 cell growth. Additional studies reveal that the inhibitors induced G2/M arrest and the (S)-enantiomer of the phenylglycine-based derivatives was a more potent inhibitor of cytosolic iPLA(2)β.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%
Supelco
Morpholine, analytical standard