Merck
Flow injection-chemiluminescence determination of dopamine using potassium permanganate and formaldehyde system.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-06-12)
Saikh Mohammad Wabaidur, Zeid Abdullah Alothman, Seikh Mafiz Alam, Sang Hak Lee
ABSTRAKT
A simple and sensitive flow injection-chemiluminescence method for the determination of dopamine has been proposed. The method is based on the enhancing effect of dopamine on the chemiluminescence emission generated by the reaction of potassium permanganate with formaldehyde in an acidic medium. The proposed procedure allows the determination of dopamine over the concentration range of 3.1×10(-8)-1.7×10(-5) mol/L and with a detection limit of 1.0×10(-8) mol/L. The linear regression equation was F=44.4912+1.07×10(9)(*)C (correlation coefficient, r(2)=0.9998). The relative standard deviation is 2.1% for the determination of 1.0×10(-8) mol/L dopamine (n=11). The method was successfully applied to the determination of dopamine in pharmaceutical preparation with satisfactory results. The recoveries were found in the range of 96.5-101.3%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium permanganate, 97%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%