Merck
Prospective randomized pilot study on the effects of two synthetic high-flux dialyzers on dialysis patient anemia.
The International journal of artificial organs (2012-06-12)
Sara M Viganò, Salvatore Di Filippo, Vincenzo La Milia, Giuseppe Pontoriero, Francesco Locatelli
ABSTRAKT
Anemia in chronic kidney disease dialysis patients is a complex syndrome involving many causes. Adequate dialysis can contribute to its correction through many mechanisms, including the removal of molecules that may inhibit erythropoiesis. The aim of this pilot study was to evaluate the effect on renal anemia of two synthetic, high-flux dialyzers (polynephron vs. high-flux polysulphone). 20 dialysis patients (11 male; mean age: 72 years) were randomly assigned and studied for 6 months. There were 2 dropouts in each group. Each patient underwent 3 hemodialysis treatments per week without any difference in dialysis prescription. At T = 0 and T = 6 (after 6 months), instantaneous plasma clearances and reduction rates of small solutes, ß2-microglobulin protein (ß2-µ); hemoglobin (Hb), and iron pattern were measured. The effect on anemia was evaluated by calculating the Erythropoesis Stimulating Agent (ESA) doses and the Erythropoietin Resistance Index (ERI). Kt/V increased between T0 and T6 in both groups. ß2-µ pre-dialysis levels significantly decreased between T0 and T6 in both dialyzer groups (p<0.001 in both groups). The Hb levels increased between T0 and T6, but significantly only for the polynephron patient group (p = 0.006 and 0.142). ESA dose did not change significantly. The ERI decreased by 22.7% between T0 and T6 in the polynephron-group and increased by 14% in the others; these changes were not significant. High-flux filters improved Hb levels, although only significantly in the polynephron group, suggesting a possible different effect. The results should be interpreted with caution and tested in an appropriately powered, large, prospective, randomized control trial.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Polysulfone, average Mn ~22,000 by MO, beads
Sigma-Aldrich
Polysulfone, average Mw ~35,000 by LS, average Mn ~16,000 by MO, pellets (Transparent)