Merck
One-step synthesis of biodegradable curcumin-derived hydrogels as potential soft tissue fillers after breast cancer surgery.
Biomacromolecules (2012-06-19)
Nava Shpaisman, Larisa Sheihet, Jared Bushman, James Winters, Joachim Kohn
ABSTRAKT
A one-step synthesis of a curcumin-derived hydrogel (curcumin content of 25-75 mol %) is reported. Curcumin is incorporated into the hydrogel backbone and cross-linked through biodegradable carbonate linkages. Curcumin as a part of the polymer backbone is protected from oxidation and degradation, while hydrogel hydrolysis results in the release of active curcumin. Nontoxic poly(ethylene glycol) and desaminotyrosyl-tyrosine ethyl ester are used to tune the hydrophilic/hydrophobic hydrogel properties. In this way, hydrogels with a wide range of physical properties including water-uptake (100-550%) and compression moduli (7-100 kPa) were obtained. Curcumin release is swelling-controlled and could be extended to 80 days. In vitro, curcumin-derived hydrogels showed selective cytotoxicity against MDA-MB-231 (IC(50) 9 μM) breast cancer cells but no cytotoxicity to noncancerous quiescent human dermal fibroblasts even at high curcumin concentrations (160 μM). One possible application of these curcumin-derived hydrogels is as soft tissue filler after surgical removal of cancerous tissue.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Sigma-Aldrich
Phosgene solution, ~20% in toluene
Sigma-Aldrich
Phosgene solution, 15 wt. % in toluene
Sigma-Aldrich
Triphosgene, reagent grade, 98%