Merck
Zonisamide for the treatment of self-inflicted dermatoses related to impulse control disorders.
Archives of dermatology (2012-06-20)
Maria José Tribó, Eduardo Alejandro Rozas-Muñoz, Fernando Gallardo, Noemí Casanova, Salvador Ros-Montalban, Antoni Bulbena, Ramon Pujol
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Zonisamide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Zonisamide sodium salt, ≥98% (HPLC), powder