Merck
Preparation of mesoporous/macroporous materials in highly concentrated emulsions based on cubic phases by a single-step method.
Langmuir : the ACS journal of surfaces and colloids (2012-06-28)
Jordi Esquena, Jérémie Nestor, Alejandro Vílchez, Kenji Aramaki, Conxita Solans
ABSTRAKT
A novel and simple single-step method for the preparation of meso/macroporous silica materials is described, which consists in templating in highly concentrated emulsions with a cubic liquid crystal in the continuous phase. Tetraethyl orthosilicate (TEOS) was solubilized in the aqueous continuous phase of highly concentrated emulsions stabilized by C(12)(EO)(8) and a PEO-PPO-PEO block copolymer nonionic surfactant, with a cubic liquid crystalline phase of the Fd3m type. The resulting silica materials were characterized by small-angle X-ray scattering, nitrogen sorption and transmission electron microscopy. The results showed that a dual pore size distribution was obtained, consisting of mesopores in the nanometer range and macropores between 1 and 5 μm. These dual meso/macroporous silicas with bimodal pore size distribution can possess specific surface areas higher than 400 m(2)/g.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Decane, analytical standard
Sigma-Aldrich
Decane, anhydrous, ≥99%
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Decane, ReagentPlus®, ≥99%
Sigma-Aldrich
Decane, ≥95%
Sigma-Aldrich
Poly(ethylene glycol-ran-propylene glycol) monobutyl ether, average Mn ~3,900