Merck
  • Selective imaging of active pharmaceutical ingredients in powdered blends with common excipients utilizing two-photon excited ultraviolet-fluorescence and ultraviolet-second order nonlinear optical imaging of chiral crystals.
Selective imaging of active pharmaceutical ingredients in powdered blends with common excipients utilizing two-photon excited ultraviolet-fluorescence and ultraviolet-second order nonlinear optical imaging of chiral crystals.
Analytical chemistry (2012-07-24)
S J Toth, J T Madden, L S Taylor, P Marsac, G J Simpson
ABSTRAKT
Second order nonlinear optical imaging of chiral crystals (SONICC) and two-photon excited fluorescence measurements [both autofluorescence and two-photon excited UV-fluorescence (TPE-UVF)] were assessed for the selective detection of APIs relative to common pharmaceutical excipients. Active pharmaceutical ingredients (APIs) compose only a small percentage of most tabulated formulations, yet the API distribution within the tablet can affect drug release and tablet stability. Complementary measurements using either UV-SONICC (266 nm detection) or TPE-UVF were shown to generate signals >50-fold more intense for a model API (griseofulvin) than those produced by common pharmaceutical excipients. The combined product of the measurements produced signals >10(4)-fold greater than the excipients studied. UV-SONICC or TPE-UVF produced greater selectivity than analogous measurements with visible-light detection, attributed to the presence of aromatic moieties within the API exhibiting strong one and two photon absorption at ~266 nm. Complementary SONICC and fluorescence measurements allowed for the sensitive detection of the three-dimensional distribution of tadalafil within a Cialis tablet to a depth of >140 μm.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%