Merck
Ca²⁺-dependent ion channels underlying spontaneous activity in insect circadian pacemaker neurons.
The European journal of neuroscience (2012-07-24)
Hongying Wei, Monika Stengl
ABSTRAKT
Electrical activity in the gamma frequency range is instrumental for temporal encoding on the millisecond scale in attentive vertebrate brains. Surprisingly, also circadian pacemaker neurons in the cockroach Rhyparobia maderae (Leucophaea maderae) employ fast spontaneous rhythmic activity in the gamma band frequency range (20-70  Hz) together with slow rhythmic activity. The ionic conductances controlling this fast spontaneous activity are still unknown. Here, Ca(2+) imaging combined with pharmacology was employed to analyse ion channels underlying spontaneous activity in dispersed circadian pacemakers of the adult accessory medulla, which controls circadian locomotor activity rhythms. Fast spontaneous Ca(2+) transients in circadian pacemakers accompany tetrodotoxin (TTX)-blockable spontaneous action potentials. In contrast to vertebrate pacemakers, the spontaneous depolarisations from rest appear to be rarely initiated via TTX-sensitive sustained Na(+) channels. Instead, they are predominantly driven by mibefradil-sensitive, low-voltage-activated Ca(2+) channels and DK-AH269-sensitive hyperpolarisation-activated, cyclic nucleotide-gated cation channels. Rhythmic depolarisations activate voltage-gated Na(+) channels and nifedipine-sensitive high-voltage-activated Ca(2+) channels. Together with Ca(2+) rises, the depolarisations open repolarising small-conductance but not large-conductance Ca(2+) -dependent K(+) channels. In contrast, we hypothesise that P/Q-type Ca(2+) channels coupled to large-conductance Ca(2+) -dependent K(+) channels are involved in input-dependent activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Mibefradil dihydrochloride hydrate, ≥98% (HPLC), powder