Merck
Performance of an organic photodiode as an optical detector and its application to fluorometric flow-immunoassay for IgA.
Talanta (2012-07-24)
Mayo Miyake, Hizuru Nakajima, Akihide Hemmi, Masayuki Yahiro, Chihaya Adachi, Nobuaki Soh, Ryoichi Ishimatsu, Koji Nakano, Katsumi Uchiyama, Toshihiko Imato
ABSTRAKT
The performance of an organic thin film photodiode (OPD), fabricated from a hetero-junction comprised of two layers of C(60) and a phthalocyanine-Cu(II) complex was evaluated by detecting the chemiluminescence generated from the reaction of luminol with horseradish peroxidase in the presence of H(2)O(2), and the fluorescence from resorufin, as an optical detector. The photocurrent of the OPD was linear with respect to the power of light from a commercial LED. The sensitivity of the OPD was sufficient for detecting chemiluminescence with a power 0.1μW/cm(2). The OPD was successfully used in a flow-immunoassay for IgA, a marker of human stress, in which a sandwich immunoassay was carried out on the microchip and the fluorescence from resorufin, produced by the enzymatic reaction, was detected. The detection limits for resorufin and IgA were 5.0μM and 16ng/mL, respectively. The photosensitivity of the OPD remained relatively constant for a minimum of one year.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Copper(II) phthalocyanine, sublimed grade, Dye content 99 %
Sigma-Aldrich
Copper(II) phthalocyanine, Dye content >99 %
Sigma-Aldrich
Copper(II) phthalocyanine, β-form, Dye content 90 %
Sigma-Aldrich
Copper(II) phthalocyanine, triple-sublimed grade, >99.95% trace metals basis